Merck
  • Hormone-sensitive lipase couples intergenerational sterol metabolism to reproductive success.
Hormone-sensitive lipase couples intergenerational sterol metabolism to reproductive success.
eLife (2021-02-05)
Christoph Heier, Oskar Knittelfelder, Harald F Hofbauer, Wolfgang Mende, Ingrid Pörnbacher, Laura Schiller, Gabriele Schoiswohl, Hao Xie, Sebastian Grönke, Andrej Shevchenko, Ronald P Kühnlein
ABSTRACT
Triacylglycerol (TG) and steryl ester (SE) lipid storage is a universal strategy to maintain organismal energy and membrane homeostasis. Cycles of building and mobilizing storage fat are fundamental in (re)distributing lipid substrates between tissues or to progress ontogenetic transitions. In this study, we show that Hormone-sensitive lipase (Hsl) specifically controls SE mobilization to initiate intergenerational sterol transfer in Drosophila melanogaster. Tissue-autonomous Hsl functions in the maternal fat body and germline coordinately prevent adult SE overstorage and maximize sterol allocation to embryos. While Hsl-deficiency is largely dispensable for normal development on sterol-rich diets, animals depend on adipocyte Hsl for optimal fecundity when dietary sterol becomes limiting. Notably, accumulation of SE but not of TG is a characteristic of Hsl-deficient cells across phyla including murine white adipocytes. In summary, we identified Hsl as an ancestral regulator of SE degradation, which improves intergenerational sterol transfer and reproductive success in flies.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Phosphatidylinositol ammonium salt solution, from soybean, ≥98.0% (TLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)