Merck
  • Structural basis for altered activity of M- and H-isozyme forms of human lactate dehydrogenase.
Structural basis for altered activity of M- and H-isozyme forms of human lactate dehydrogenase.
Proteins (2001-03-29)
J A Read, V J Winter, C M Eszes, R B Sessions, R L Brady
ABSTRACT
Lactate dehydrogenase (LDH) interconverts pyruvate and lactate with concomitant interconversion of NADH and NAD(+). Although crystal structures of a variety of LDH have previously been described, a notable absence has been any of the three known human forms of this glycolytic enzyme. We have now determined the crystal structures of two isoforms of human LDH-the M form, predominantly found in muscle; and the H form, found mainly in cardiac muscle. Both structures have been crystallized as ternary complexes in the presence of the NADH cofactor and oxamate, a substrate-like inhibitor. Although each of these isoforms has different kinetic properties, the domain structure, subunit association, and active-site regions are indistinguishable between the two structures. The pK(a) that governs the K(M) for pyruvate for the two isozymes is found to differ by about 0.94 pH units, consistent with variation in pK(a) of the active-site histidine. The close similarity of these crystal structures suggests the distinctive activity of these enzyme isoforms is likely to result directly from variation of charged surface residues peripheral to the active site, a hypothesis supported by electrostatic calculations based on each structure. Proteins 2001;43:175-185.
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein