Merck
  • Statins affect cancer cell plasticity with distinct consequences for tumor progression and metastasis.
Statins affect cancer cell plasticity with distinct consequences for tumor progression and metastasis.
Cell reports (2021-11-25)
Madeleine Dorsch, Manuela Kowalczyk, Mélanie Planque, Geronimo Heilmann, Sebastian Urban, Philip Dujardin, Jan Forster, Kristina Ueffing, Silke Nothdurft, Sebastian Oeck, Annika Paul, Sven T Liffers, Farnusch Kaschani, Markus Kaiser, Alexander Schramm, Jens T Siveke, Monte M Winslow, Sarah-Maria Fendt, Perihan Nalbant, Barbara M Grüner
ABSTRACT
Statins are among the most commonly prescribed drugs, and around every fourth person above the age of 40 is on statin medication. Therefore, it is of utmost clinical importance to understand the effect of statins on cancer cell plasticity and its consequences to not only patients with cancer but also patients who are on statins. Here, we find that statins induce a partial epithelial-to-mesenchymal transition (EMT) phenotype in cancer cells of solid tumors. Using a comprehensive STRING network analysis of transcriptome, proteome, and phosphoproteome data combined with multiple mechanistic in vitro and functional in vivo analyses, we demonstrate that statins reduce cellular plasticity by enforcing a mesenchymal-like cell state that increases metastatic seeding ability on one side but reduces the formation of (secondary) tumors on the other due to heterogeneous treatment responses. Taken together, we provide a thorough mechanistic overview of the consequences of statin use for each step of cancer development, progression, and metastasis.
Sigma-Aldrich
Tipifarnib, ≥98% (HPLC)
Sigma-Aldrich
Dasatinib, ≥98% (HPLC)
Sigma-Aldrich
Zoledronic acid monohydrate, ≥98% (HPLC)