Merck
Robust olfactory responses in the absence of odorant binding proteins.
eLife (2019-10-28)
Shuke Xiao, Jennifer S Sun, John R Carlson
ABSTRACT
Odorant binding proteins (Obps) are expressed at extremely high levels in the antennae of insects, and have long been believed essential for carrying hydrophobic odorants to odor receptors. Previously we found that when one functional type of olfactory sensillum in Drosophila was depleted of its sole abundant Obp, it retained a robust olfactory response (Larter et al., 2016). Here we have deleted all the Obp genes that are abundantly expressed in the antennal basiconic sensilla. All of six tested sensillum types responded robustly to odors of widely diverse chemical or temporal structure. One mutant gave a greater physiological and behavioral response to an odorant that affects oviposition. Our results support a model in which many sensilla can respond to odorants in the absence of Obps, and many Obps are not essential for olfactory response, but that some Obps can modulate olfactory physiology and the behavior that it drives.
Sigma-Aldrich
Linoleic acid, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-Geosmin solution, 2 mg/mL in methanol, ≥97%
Pricing and availability is not currently available.