Merck
  • Proteomic analysis of decidua in patients with recurrent pregnancy loss (RPL) reveals mitochondrial oxidative stress dysfunction.
Proteomic analysis of decidua in patients with recurrent pregnancy loss (RPL) reveals mitochondrial oxidative stress dysfunction.
Clinical proteomics (2021-02-24)
Xiang-Jie Yin, Wei Hong, Fu-Ju Tian, Xiao-Cui Li
ABSTRACT
Pregnancy is a complicated physiological process. The multifaceted regulation of maternal-fetal interface is of great importance for maintaining normal pregnancy and avoiding fetal rejection and secondary abortion. Previous studies have focused on the clinical features or pathological biomarkers of fetal rejection and abortion. However, no significant breakthrough has been made. Therefore, it is important to understand the molecular mechanisms of recurrent pregnancy loss (RPL) to identify potential therapeutic strategies. The aim of this study was to investigate the pathogenesis of RPL. In this study, Relative and absolute quantitation (iTRAQ) technology integrated with liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis was used to identify differentially expressed proteins in decidual from RPL patients and matched normal controls. Further, Molecules NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3 (ndufb3) and cyclooxygenase-2 (COX-2) were validated by immunohistochemistry (IHC), Western blotting, CCK8 and mitochondrial red fluorescent probe (Mito-Tracker Red CMXRos). A total of 456 proteins reached the threshold of a 1.5-fold change were identified for further bioinformatics analysis. Upon mapping the differentially expressed proteins using the Kyoto Encyclopedia of Genes and Genomes (KEGG) pathways database, iTRAQ results were confirmed by assessing NDUFB3 and COX-2 protein levels in specimens of decidual tissue by Western blotting. Our study indicates that the level of COX-2 and NDUFB3 were significantly increased in decidual cell from RPL patients. Overexpression of NDUFB3 inhibited cell vitality and oxidative stress of decimal cell. Further, our found that overexpression NDUFBD3 in decidual cell decreased the mitochondrial membrane potential expression levels. These results suggest that NDUFB3 might play an important role in promote the pathological process of RPL. This comprehensive analysis of RPL proteomics reveals novel candidate: NDUFB3, which could be further investigated for explanation of the pathological mechanism of RPL.