Merck
  • Single-cell analysis reveals urothelial cell heterogeneity and regenerative cues following cyclophosphamide-induced bladder injury.
Single-cell analysis reveals urothelial cell heterogeneity and regenerative cues following cyclophosphamide-induced bladder injury.
Cell death & disease (2021-05-07)
Xiaomu Cheng, Huadong Lai, Wenqin Luo, Man Zhang, Juju Miao, Weichen Song, Shunpeng Xing, Jia Wang, Wei-Qiang Gao
ABSTRACT
Cyclophosphamide is a commonly used chemotherapeutic drug to treat cancer with side effects that trigger bladder injury and hemorrhagic cystitis. Although previous studies have demonstrated that certain cell subsets and communications are activated to drive the repair and regeneration of bladder, it is not well understood how distinct bladder cell subsets function synergistically in this process. Here, we used droplet-based single-cell RNA sequencing (scRNA-seq) to profile the cell types within the murine bladder mucous layer under normal and injured conditions. Our analysis showed that superficial cells are directly repaired by cycling intermediate cells. We further identified two resident mesenchymal lineages (Acta2+ myofibroblasts and Cd34+ fibroblasts). The delineation of cell-cell communications revealed that Acta2+ myofibroblasts upregulated Fgf7 expression during acute injury, which activated Fgfr signaling in progenitor cells within the basal/intermediate layers to promote urothelial cell growth and repair. Overall, our study contributes to a more comprehensive understanding of the cellular dynamics during cyclophosphamide-induced bladder injury and may help identify important niche factors contributing to the regeneration of injured bladders.
Sigma-Aldrich
DNase I, Amplification Grade