Merck
A tamoxifen receptor within a voltage-gated sodium channel.
Molecular cell (2021-01-28)
Altin Sula, David Hollingworth, Leo C T Ng, Megan Larmore, Paul G DeCaen, B A Wallace
ABSTRACT
Voltage-gated sodium channels are targets for many analgesic and antiepileptic drugs whose therapeutic mechanisms and binding sites have been well characterized. We describe the identification of a previously unidentified receptor site within the NavMs voltage-gated sodium channel. Tamoxifen, an estrogen receptor modulator, and its primary and secondary metabolic products bind at the intracellular exit of the channel, which is a site that is distinct from other previously characterized sodium channel drug sites. These compounds inhibit NavMs and human sodium channels with similar potencies and prevent sodium conductance by delaying channel recovery from the inactivated state. This study therefore not only describes the structure and pharmacology of a site that could be leveraged for the development of new drugs for the treatment of sodium channelopathies but may also have important implications for off-target health effects of this widely used therapeutic drug.
Sigma-Aldrich
Poly(ethylene glycol), average Mn 400
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
N-Desmethyltamoxifen HCl, ≥98% (HPLC), solid
Sigma-Aldrich
(E/Z)-Endoxifen Hydrochloride Hydrate, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride dihydrate, ≥99.0%