Merck
  • Opposing Wnt signals regulate cervical squamocolumnar homeostasis and emergence of metaplasia.
Opposing Wnt signals regulate cervical squamocolumnar homeostasis and emergence of metaplasia.
Nature cell biology (2021-01-20)
Cindrilla Chumduri, Rajendra Kumar Gurumurthy, Hilmar Berger, Oliver Dietrich, Naveen Kumar, Stefanie Koster, Volker Brinkmann, Kirstin Hoffmann, Marina Drabkina, Panagiota Arampatzi, Dajung Son, Uwe Klemm, Hans-Joachim Mollenkopf, Hermann Herbst, Mandy Mangler, Jörg Vogel, Antoine-Emmanuel Saliba, Thomas F Meyer
ABSTRACT
The transition zones of the squamous and columnar epithelia constitute hotspots for the emergence of cancer, often preceded by metaplasia, in which one epithelial type is replaced by another. It remains unclear how the epithelial spatial organization is maintained and how the transition zone niche is remodelled during metaplasia. Here we used single-cell RNA sequencing to characterize epithelial subpopulations and the underlying stromal compartment of endo- and ectocervix, encompassing the transition zone. Mouse lineage tracing, organoid culture and single-molecule RNA in situ hybridizations revealed that the two epithelia derive from separate cervix-resident lineage-specific stem cell populations regulated by opposing Wnt signals from the stroma. Using a mouse model of cervical metaplasia, we further show that the endocervical stroma undergoes remodelling and increases expression of the Wnt inhibitor Dickkopf-2 (DKK2), promoting the outgrowth of ectocervical stem cells. Our data indicate that homeostasis at the transition zone results from divergent stromal signals, driving the differential proliferation of resident epithelial lineages.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)