Merck
  • Coding and non-coding roles of MOCCI (C15ORF48) coordinate to regulate host inflammation and immunity.
Coding and non-coding roles of MOCCI (C15ORF48) coordinate to regulate host inflammation and immunity.
Nature communications (2021-04-11)
Cheryl Q E Lee, Baptiste Kerouanton, Sonia Chothani, Shan Zhang, Ying Chen, Chinmay Kumar Mantri, Daniella Helena Hock, Radiance Lim, Rhea Nadkarni, Vinh Thang Huynh, Daryl Lim, Wei Leong Chew, Franklin L Zhong, David Arthur Stroud, Sebastian Schafer, Vinay Tergaonkar, Ashley L St John, Owen J L Rackham, Lena Ho
ABSTRACT
Mito-SEPs are small open reading frame-encoded peptides that localize to the mitochondria to regulate metabolism. Motivated by an intriguing negative association between mito-SEPs and inflammation, here we screen for mito-SEPs that modify inflammatory outcomes and report a mito-SEP named "Modulator of cytochrome C oxidase during Inflammation" (MOCCI) that is upregulated during inflammation and infection to promote host-protective resolution. MOCCI, a paralog of the NDUFA4 subunit of cytochrome C oxidase (Complex IV), replaces NDUFA4 in Complex IV during inflammation to lower mitochondrial membrane potential and reduce ROS production, leading to cyto-protection and dampened immune response. The MOCCI transcript also generates miR-147b, which targets the NDUFA4 mRNA with similar immune dampening effects as MOCCI, but simultaneously enhances RIG-I/MDA-5-mediated viral immunity. Our work uncovers a dual-component pleiotropic regulation of host inflammation and immunity by MOCCI (C15ORF48) for safeguarding the host during infection and inflammation.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Adenosine 5′-diphosphate monopotassium salt dihydrate, bacterial, ≥95%, powder
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Acetyl coenzyme A lithium salt, ≥93% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Lactate Dehydrogenase Activity Assay Kit, sufficient for 500 colorimetric tests
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks