Merck
  • IgM and IgA augmented autoantibody signatures improve early-stage detection of colorectal cancer prior to nodal and distant spread.
IgM and IgA augmented autoantibody signatures improve early-stage detection of colorectal cancer prior to nodal and distant spread.
Clinical & translational immunology (2021-10-05)
Md Saiful Islam Roney, Catharine Lanagan, Yong Hua Sheng, Karen Lawler, Christopher Schmidt, Nam-Trung Nguyen, Jakob Begun, Gregor Stefan Kijanka
ABSTRACT
Tumor-associated autoantibodies (AAbs) in individuals with cancer can precede clinical diagnosis by several months to years. The objective of this study was to determine whether the primary immune response in form of IgM and gut mucosa-associated IgA can aid IgG AAbs in the detection of early-stage colorectal cancer (CRC). We developed a novel protein array comprising 492 antigens seropositive in CRC. The array was used to profile IgG, IgM and IgA antibody signatures in 99 CRC patients and 99 sex- and age-matched non-cancer controls. A receiver operating curve (ROC), Kaplan-Meier survival analysis and univariate and multivariate Cox regression analyses were conducted. We identified a panel of 16 multi-isotype AAbs with a cumulative sensitivity of 91% and specificity of 74% (AUC 0.90, 95% CI: 0.850-0.940) across all CRC stages. IgM and IgG isotypes were conversely associated with disease stage with IgM contributing significantly to improved stage I and II sensitivity of 96% at 78% specificity (AUC 0.928, 95% CI: 0.884-0.973). A single identified IgA AAb reached an overall sensitivity of 5% at 99% specificity (AUC 0.520, 95% CI: 0.440-0.601) balanced across all CRC stages. Kaplan-Meier analysis revealed that se33-1 (ZNF638) IgG AAbs were associated with reduced 5-year overall survival (log-rank test, P = 0.012), whereas cumulative IgM isotype signatures were associated with improved 5-year overall survival (log-rank test, P = 0.024). IgM AAbs are associated with early-stage colorectal cancer. Combining IgG, IgM and IgA AAbs is a novel strategy to improve early diagnosis of cancers.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution