Merck
  • NF-κB-inducing kinase maintains T cell metabolic fitness in antitumor immunity.
NF-κB-inducing kinase maintains T cell metabolic fitness in antitumor immunity.
Nature immunology (2021-01-06)
Meidi Gu, Xiaofei Zhou, Jee Hyung Sohn, Lele Zhu, Zuliang Jie, Jin-Young Yang, Xiaofeng Zheng, Xiaoping Xie, Jie Yang, Yaoyao Shi, Hans D Brightbill, Jae Bum Kim, Jing Wang, Xuhong Cheng, Shao-Cong Sun
ABSTRACT
Metabolic reprograming toward aerobic glycolysis is a pivotal mechanism shaping immune responses. Here we show that deficiency in NF-κB-inducing kinase (NIK) impairs glycolysis induction, rendering CD8+ effector T cells hypofunctional in the tumor microenvironment. Conversely, ectopic expression of NIK promotes CD8+ T cell metabolism and effector function, thereby profoundly enhancing antitumor immunity and improving the efficacy of T cell adoptive therapy. NIK regulates T cell metabolism via a NF-κB-independent mechanism that involves stabilization of hexokinase 2 (HK2), a rate-limiting enzyme of the glycolytic pathway. NIK prevents autophagic degradation of HK2 through controlling cellular reactive oxygen species levels, which in turn involves modulation of glucose-6-phosphate dehydrogenase (G6PD), an enzyme that mediates production of the antioxidant NADPH. We show that the G6PD-NADPH redox system is important for HK2 stability and metabolism in activated T cells. These findings establish NIK as a pivotal regulator of T cell metabolism and highlight a post-translational mechanism of metabolic regulation.