Merck
  • Toxicity and Local Tolerance of COVID- eVax, a Plasmid DNA Vaccine for SARS-CoV-2, Delivered by Electroporation.
Toxicity and Local Tolerance of COVID- eVax, a Plasmid DNA Vaccine for SARS-CoV-2, Delivered by Electroporation.
Toxicologic pathology (2021-09-09)
Yuval Ramot, Gianfranco Caselli, Luigi Aurisicchio, Isabella Andreini, Emanuele Marra, Laura Luberto, Daniela Stoppoloni, Maria Lucrezia Pacello, Laura Monetini, Abraham Nyska
ABSTRACT
COVID-19 is a rapidly spreading disease, posing a huge hazard to global health. The plasmid vaccine pTK1A-TPA-SpikeA (named COVID-eVax) encodes the severe acute respiratory syndrome coronavirus 2 S protein receptor-binding domain, developed for intramuscular injection followed by electroporation (EP). The aim of this study was to assess the systemic toxicity and local tolerance of COVID-eVax delivered intramuscularly followed by EP in Sprague Dawley (SD) rats. The animals were killed 2 days and 4 weeks after the last injection (30-day and 57-day, respectively). No mortality was observed, and no signs of toxicity were evident, including injection site reactions. A lasting and specific immune response was observed in all treated animals, confirming the relevance of the rat as a toxicological model for this vaccine. Histopathological evaluation revealed muscle fiber necrosis associated with subchronic inflammation at the injection sites (at the 30-day time point), with a clear trend for recovery at the 57-day time point, which is expected following EP, and considered a desirable effect to mount the immune response against the target antigen. In conclusion, the intramuscular EP-assisted DNA vaccine, COVID-eVax showed an excellent safety profile in SD rats under these experimental conditions and supports its further development for use in humans.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA, ready to use solution