Merck
  • Retrospective cell lineage reconstruction in humans by using short tandem repeats.
Retrospective cell lineage reconstruction in humans by using short tandem repeats.
Cell reports methods (2021-08-04)
Liming Tao, Ofir Raz, Zipora Marx, Manjusha S Ghosh, Sandra Huber, Julia Greindl-Junghans, Tamir Biezuner, Shiran Amir, Lilach Milo, Rivka Adar, Ron Levy, Amos Onn, Noa Chapal-Ilani, Veronika Berman, Asaf Ben Arie, Guy Rom, Barak Oron, Ruth Halaban, Zbigniew T Czyz, Melanie Werner-Klein, Christoph A Klein, Ehud Shapiro
ABSTRACT
Cell lineage analysis aims to uncover the developmental history of an organism back to its cell of origin. Recently, novel in vivo methods utilizing genome editing enabled important insights into the cell lineages of animals. In contrast, human cell lineage remains restricted to retrospective approaches, which still lack resolution and cost-efficient solutions. Here, we demonstrate a scalable platform based on short tandem repeats targeted by duplex molecular inversion probes. With this human cell lineage tracing method, we accurately reproduced a known lineage of DU145 cells and reconstructed lineages of healthy and metastatic single cells from a melanoma patient who matched the anatomical reference while adding further refinements. This platform allowed us to faithfully recapitulate lineages of developmental tissue formation in healthy cells. In summary, our lineage discovery platform can profile informative somatic mutations efficiently and provides solid lineage reconstructions even in challenging low-mutation-rate healthy single cells.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Phenol – chloroform – isoamyl alcohol mixture, BioUltra, for molecular biology, 25:24:1
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Pricing and availability is not currently available. Contact Technical Service