Merck
  • Potent preclinical sensitivity to imipridone-based combination therapies in oncohistone H3K27M-mutant diffuse intrinsic pontine glioma is associated with induction of the integrated stress response, TRAIL death receptor DR5, reduced ClpX and apoptosis.
Potent preclinical sensitivity to imipridone-based combination therapies in oncohistone H3K27M-mutant diffuse intrinsic pontine glioma is associated with induction of the integrated stress response, TRAIL death receptor DR5, reduced ClpX and apoptosis.
American journal of cancer research (2021-10-19)
Robyn Borsuk, Lanlan Zhou, Wen-I Chang, Yiqun Zhang, Aditi Sharma, Varun V Prabhu, Nikos Tapinos, Rishi R Lulla, Wafik S El-Deiry
ABSTRACT
The H3K27M oncohistone mutation, identified in approximately 80% of diffuse intrinsic pontine gliomas (DIPG), is a potential target for therapy. Imipridone ONC201/TIC10 (TRAIL-Inducing Compound #10) induces apoptosis of cancer cells, and has clinical efficacy against H3K27M-mutant DIPG. We demonstrate synergy between ONC201, ONC206 and ONC212, and targeted therapies with known preclinical activity against DIPG. We hypothesized that imipridone combinations with HDAC or proteasome inhibitors may be superior to single agent ONC201 treatment in H3K27M mutant DIPG. Six patient-derived DIPG cell lines (SU-DIPG-IV, SU-DIPG-13, SU-DIPG-25, SU-DIPG-27, SU-DIPG-29, SU-DIPG-36) were exposed to imipridones alone or combinations with histone de-acetylase inhibitors [HDACi], marizomib, etoposide, and temozolomide. Dose-dependent response to imipridones was observed in DIPG cells with half-maximal inhibitory concentration (IC50) of 1.46 µM, 0.11 µM, and 0.03 µM, for ONC201, ONC206, and ONC212, respectively. Upon treatment with the imipridones, DIPG cell lines engaged CLpP/CLPX, the integrated stress response with ATF4 activation, and TRAIL death receptor 5 (DR5) induction. Strong synergy was identified between ONC201 and HDACi panobinostat (combination index [CI] 0.01), romidepsin (CI 0.08) and proteasome inhibitor marizomib (CI 0.19). Synergy was demonstrated between ONC201 and etoposide (CI 0.54), although to a lesser degree than with panobinostat, romidepsin, and marizomib. ONC206 and ONC212 showed similar synergistic effects with panobinostat, romidepsin, and marizomib. Induction of apoptosis was demonstrated with imipridones and panobinostat or romidepsin combinations. Our results suggest increased sensitivity of H3K27M-mutant DIPG cell lines to second generation imipridone therapies, as compared to ONC201. Additionally, there is synergistic cell death with combination of imipridones and panobinostat, romidepsin, or marizomib, which may be further tested in vivo and in clinical trials.