- Chicken IgY: utilizing the evolutionary difference.
Chicken IgY: utilizing the evolutionary difference.
Comparative immunology, microbiology and infectious diseases (1990-01-01)
A Larsson, J Sjöquist
PMID2076606
ABSTRACT
Chicken antibodies offer advantages over traditional ones (e.g. rabbit antibodies) due to the evolutionary difference between these immunoglobulins. Chicken give better antibody response against conserved mammalian antigens, they do not react with rheumatoid factors, bacterial or mammalian Fc receptors and can reduce background reactions due to cross-reactivity of anti-IgG antibodies.