Merck
  • bFGF and SDF-1α Improve In Vivo Performance of VEGF-Incorporating Small-Diameter Vascular Grafts.
bFGF and SDF-1α Improve In Vivo Performance of VEGF-Incorporating Small-Diameter Vascular Grafts.
Pharmaceuticals (Basel, Switzerland) (2021-04-04)
Larisa Antonova, Anton Kutikhin, Viktoriia Sevostianova, Elena Velikanova, Vera Matveeva, Tatiana Glushkova, Andrey Mironov, Evgeniya Krivkina, Amin Shabaev, Evgeniya Senokosova, Leonid Barbarash
ABSTRACT
Tissue-engineered vascular grafts are widely tested as a promising substitute for both arterial bypass and replacement surgery. We previously demonstrated that incorporation of VEGF into electrospun tubular scaffolds from poly(3-hydroxybutyrate-co-3-hydroxyvalerate)/poly(ε-caprolactone) enhances formation of an endothelial cell monolayer. However, an overdose of VEGF can induce tumor-like vasculature; thereby, other bioactive factors are needed to support VEGF-driven endothelialization and successful recruitment of smooth muscle cells. Utilizing emulsion electrospinning, we fabricated one-layer vascular grafts with either VEGF, bFGF, or SDF-1α, and two-layer vascular grafts with VEGF incorporated into the inner layer and bFGF and SDF-1α incorporated into the outer layer with the following structural evaluation, tensile testing, and in vivo testing using a rat abdominal aorta replacement model. The latter graft prototype showed higher primary patency rate. We found that the two-layer structure improved surface topography and mechanical properties of the grafts. Further, the combination of bFGF, SDF-1α, and VEGF improved endothelialization compared with VEGF alone, while bFGF induced a rapid formation of a smooth muscle cell layer. Taken together, these findings show that the two-layer structure and incorporation of bFGF and SDF-1α into the vascular grafts in combination with VEGF provide a higher primary patency and therefore improved in vivo performance.
Sigma-Aldrich
VEGF165 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Polycaprolactone, average Mn 80,000