Merck
  • Polyamine metabolism is a central determinant of helper T cell lineage fidelity.
Polyamine metabolism is a central determinant of helper T cell lineage fidelity.
Cell (2021-07-04)
Daniel J Puleston, Francesc Baixauli, David E Sanin, Joy Edwards-Hicks, Matteo Villa, Agnieszka M Kabat, Marcin M Kamiński, Michal Stanckzak, Hauke J Weiss, Katarzyna M Grzes, Klara Piletic, Cameron S Field, Mauro Corrado, Fabian Haessler, Chao Wang, Yaarub Musa, Lena Schimmelpfennig, Lea Flachsmann, Gerhard Mittler, Nir Yosef, Vijay K Kuchroo, Joerg M Buescher, Stefan Balabanov, Edward J Pearce, Douglas R Green, Erika L Pearce
ABSTRACT
Polyamine synthesis represents one of the most profound metabolic changes during T cell activation, but the biological implications of this are scarcely known. Here, we show that polyamine metabolism is a fundamental process governing the ability of CD4+ helper T cells (TH) to polarize into different functional fates. Deficiency in ornithine decarboxylase, a crucial enzyme for polyamine synthesis, results in a severe failure of CD4+ T cells to adopt correct subset specification, underscored by ectopic expression of multiple cytokines and lineage-defining transcription factors across TH cell subsets. Polyamines control TH differentiation by providing substrates for deoxyhypusine synthase, which synthesizes the amino acid hypusine, and mice in which T cells are deficient for hypusine develop severe intestinal inflammatory disease. Polyamine-hypusine deficiency caused widespread epigenetic remodeling driven by alterations in histone acetylation and a re-wired tricarboxylic acid (TCA) cycle. Thus, polyamine metabolism is critical for maintaining the epigenome to focus TH cell subset fidelity.
Product Description
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture