Merck
  • Preparation and characterization of toxic Abeta aggregates for structural and functional studies in Alzheimer's disease research.
Preparation and characterization of toxic Abeta aggregates for structural and functional studies in Alzheimer's disease research.
Nature protocols (2010-06-12)
Asad Jan, Dean M Hartley, Hilal A Lashuel
ABSTRACT
The amyloid cascade hypothesis, supported by strong evidence from genetics, pathology and studies using animal models, implicates amyloid-beta (Abeta) oligomerization and fibrillogenesis as central causative events in the pathogenesis of Alzheimer's disease (AD). Today, significant efforts in academia, biotechnology and the pharmaceutical industry are devoted to identifying the mechanisms by which the process of Abeta aggregation contributes to neurodegeneration in AD and to the identity of the toxic Abeta species. In this paper, we describe methods and detailed protocols for reproducibly preparing Abeta aggregates of defined size distribution and morphology, including monomers, protofibrils and fibrils, using size exclusion chromatography. In addition, we describe detailed biophysical procedures for elucidating the structural features, aggregation kinetics and toxic properties of the different Abeta aggregation states, with special emphasis on protofibrillar intermediates. The information provided by this approach allows for consistent correlation between the properties of the aggregates and their toxicity toward primary neurons and/or cell lines. A better understanding of the molecular and structural basis of Abeta aggregation and toxicity is crucial for the development of effective strategies aimed at prevention and/or treatment of AD. Furthermore, the identification of specific aggregation states, which correlate with neurodegeneration in AD, could lead to the development of diagnostic tools to detect and monitor disease progression. The procedures described can be performed in as little as 1 day, or may take longer, depending on the exact toxicity assays used.
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trizma® hydrochloride solution, Molecular Biology, 1 M, BioReagent
Sigma-Aldrich
Anti-Neurofilament M (145 kDa) Antibody, CT, serum, Chemicon®