Merck
  • Global chromatin relabeling accompanies spatial inversion of chromatin in rod photoreceptors.
Global chromatin relabeling accompanies spatial inversion of chromatin in rod photoreceptors.
Science advances (2021-09-25)
Cheryl L Smith, Yemin Lan, Rajan Jain, Jonathan A Epstein, Andrey Poleshko
ABSTRACT
[Figure: see text].
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^6 units/ml
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
PD 0325901, ≥98% (HPLC)
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)