Merck
  • Circulating Levels of Bone Markers after Short-Term Intense Training with Increased Dairy Consumption in Adolescent Female Athletes.
Circulating Levels of Bone Markers after Short-Term Intense Training with Increased Dairy Consumption in Adolescent Female Athletes.
Children (Basel, Switzerland) (2021-11-28)
Panagiota Klentrou, Katherine McKee, Brandon J McKinlay, Nigel Kurgan, Brian D Roy, Bareket Falk
ABSTRACT
Thirteen female adolescent soccer players (14.3 ± 1.3 years) participated in a cross-over, double-blind trial examining the effects of Greek yogurt (GY) consumption on bone biomarkers during 5 days of intense soccer training. The study took place over two intervention weeks, which consisted of a pre-training assessment day, 5 training days, and a post-training assessment day. Participants completed the GY condition and a carbohydrate isocaloric placebo control pudding condition (CHO) in random order, 4 weeks apart. Morning, fasted, resting blood samples were collected pre- and post-training in each condition. Total osteocalcin (tOC), undercarboxylated osteocalcin (unOC), C-terminal telopeptide of type 1 collagen (CTX), osteoprotegerin (OPG), and receptor activator nuclear factor kappa-β ligand (RANKL) were measured in serum. The results showed no effects for time (pre- to post-training) or condition, and no interaction for tOC, CTX, OPG, RANKL, and the OPG/RANKL ratio. A time-by-condition interaction (p = 0.011) was observed in unOC, reflecting a post-training decrease in the GY, but not the CHO condition (-26% vs. -3%, respectively). However, relative unOC (% of tOC) decreased post-training (-16%), with no differences between conditions. These findings suggest that short-term high-impact intense training had no direct catabolic impact on bone metabolism, with GY adding no benefit beyond that of the isocaloric CHO control pudding.