Merck
  • Presynaptic cholinergic modulators as potent cognition enhancers and analgesic drugs. 1. Tropic and 2-phenylpropionic acid esters.
Presynaptic cholinergic modulators as potent cognition enhancers and analgesic drugs. 1. Tropic and 2-phenylpropionic acid esters.
Journal of medicinal chemistry (1994-05-27)
F Gualtieri, G Conti, S Dei, M P Giovannoni, F Nannucci, M N Romanelli, S Scapecchi, E Teodori, L Fanfani, C Ghelardini
ABSTRACT
Previous studies have shown that (R)-(+)-hyoscyamine has analgesic activity as a consequence of increased ACh release following antagonism of central muscarinic autoreceptors. Since the enhancement of central cholinergic transmission could be beneficial for cognitive disorders, we manipulated (R)-(+)-hyoscyamine, synthesizing several derivatives of tropic and 2-phenylpropionic acids, with the aim of obtaining drugs which are able to increase ACh release and consequently to show analgesic and nootropic activities. The results showed that several new compounds are indeed potent analgesics (with an analgesic efficacy comparable to that of morphine) and that the most potent one ((+/-)-19, PG9) also has remarkable cognition-enhancing properties. Our study confirmed that the mechanism of action involves ACh release even if it is still unclear whether only muscarinic autoreceptors or, also, heteroreceptors are involved.