Merck
  • Integrating pheromonal and spatial information in the amygdalo-hippocampal network.
Integrating pheromonal and spatial information in the amygdalo-hippocampal network.
Nature communications (2021-09-08)
María Villafranca-Faus, Manuel Esteban Vila-Martín, Daniel Esteve, Esteban Merino, Anna Teruel-Sanchis, Ana Cervera-Ferri, Joana Martínez-Ricós, Ana Lloret, Enrique Lanuza, Vicent Teruel-Martí
ABSTRACT
Vomeronasal information is critical in mice for territorial behavior. Consequently, learning the territorial spatial structure should incorporate the vomeronasal signals indicating individual identity into the hippocampal cognitive map. In this work we show in mice that navigating a virtual environment induces synchronic activity, with causality in both directionalities, between the vomeronasal amygdala and the dorsal CA1 of the hippocampus in the theta frequency range. The detection of urine stimuli induces synaptic plasticity in the vomeronasal pathway and the dorsal hippocampus, even in animals with experimentally induced anosmia. In the dorsal hippocampus, this plasticity is associated with the overexpression of pAKT and pGSK3β. An amygdalo-entorhino-hippocampal circuit likely underlies this effect of pheromonal information on hippocampal learning. This circuit likely constitutes the neural substrate of territorial behavior in mice, and it allows the integration of social and spatial information.
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-GSK3β (Ser9) Antibody, clone 2D3, clone 2D3, Upstate®, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Anti-Fluorescent Gold Antibody, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution