Merck
  • Beta-galactoside transport in bacterial membrane preparations: energy coupling via membrane-bounded D-lactic dehydrogenase.
Beta-galactoside transport in bacterial membrane preparations: energy coupling via membrane-bounded D-lactic dehydrogenase.
Proceedings of the National Academy of Sciences of the United States of America (1970-08-01)
E M Barnes, H R Kaback
ABSTRACT
The transport of beta-galactosides by isolated membrane preparations from Escherichia coli strains containing a functional y gene is markedly stimulated by the conversion of D-lactate to pyruvate. The addition of D-lactate to these membrane preparations produces a 19-fold increase in the initial rate of uptake and a 10-fold stimulation of the steady-state level of intramembranal lactose or thiomethylgalactoside. Succinate, DL-alpha-hydroxybutyrate, and L-lactate partially replace D-lactate, but are much less effective; ATP and P-enolpyruvate, in addition to a number of other metabolites and cofactors, do not stimulate lactose transport by the vesicles. Lactose uptake by the membrane preparations in the presence of D-lactate requires oxygen, and is blocked by electron transport inhibitors and proton conductors; however, uptake is not significantly inhibited by high concentrations of arsenate or oligomycin. Furthermore, the P-enolpyruvate-P-transferase system is not involved in beta-galactoside transport by the E. coli membrane vesicles. The findings indicate that the beta-galactoside uptake system is coupled to the membrane-bound D-lactic dehydrogenase via an electron transport chain but does not involve oxidative phosphorylation.
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid