Merck
  • Unbiased identification of novel transcription factors in striatal compartmentation and striosome maturation.
Unbiased identification of novel transcription factors in striatal compartmentation and striosome maturation.
eLife (2021-10-06)
Maria-Daniela Cirnaru, Sicheng Song, Kizito-Tshitoko Tshilenge, Chuhyon Corwin, Justyna Mleczko, Carlos Galicia Aguirre, Houda Benlhabib, Jaroslav Bendl, Pasha Apontes, John Fullard, Jordi Creus-Muncunill, Azadeh Reyahi, Ali M Nik, Peter Carlsson, Panos Roussos, Sean D Mooney, Lisa M Ellerby, Michelle E Ehrlich
ABSTRACT
Many diseases are linked to dysregulation of the striatum. Striatal function depends on neuronal compartmentation into striosomes and matrix. Striatal projection neurons are GABAergic medium spiny neurons (MSNs), subtyped by selective expression of receptors, neuropeptides, and other gene families. Neurogenesis of the striosome and matrix occurs in separate waves, but the factors regulating compartmentation and neuronal differentiation are largely unidentified. We performed RNA- and ATAC-seq on sorted striosome and matrix cells at postnatal day 3, using the Nr4a1-EGFP striosome reporter mouse. Focusing on the striosome, we validated the localization and/or role of Irx1, Foxf2, Olig2, and Stat1/2 in the developing striosome and the in vivo enhancer function of a striosome-specific open chromatin region 4.4 Kb downstream of Olig2. These data provide novel tools to dissect and manipulate the networks regulating MSN compartmentation and differentiation, including in human iPSC-derived striatal neurons for disease modeling and drug discovery.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, Molecular Biology
Sigma-Aldrich
Anti-Opioid Receptor Antibody, µ, pain, serum, Chemicon®
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, Alexa Fluor488 Conjugate | MABN50A4, clone 211F1.1, from mouse, ALEXA FLUOR 488
Supelco
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid, pharmaceutical impurity standard
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, ≥10 units/mg protein