Merck
  • Chromatin 3D interaction analysis of the STARD10 locus unveils FCHSD2 as a regulator of insulin secretion.
Chromatin 3D interaction analysis of the STARD10 locus unveils FCHSD2 as a regulator of insulin secretion.
Cell reports (2021-02-04)
Ming Hu, Inês Cebola, Gaelle Carrat, Shuying Jiang, Sameena Nawaz, Amna Khamis, Mickaël Canouil, Philippe Froguel, Anke Schulte, Michele Solimena, Mark Ibberson, Piero Marchetti, Fabian L Cardenas-Diaz, Paul J Gadue, Benoit Hastoy, Leonardo Almeida-Souza, Harvey McMahon, Guy A Rutter
ABSTRACT
Using chromatin conformation capture, we show that an enhancer cluster in the STARD10 type 2 diabetes (T2D) locus forms a defined 3-dimensional (3D) chromatin domain. A 4.1-kb region within this locus, carrying 5 T2D-associated variants, physically interacts with CTCF-binding regions and with an enhancer possessing strong transcriptional activity. Analysis of human islet 3D chromatin interaction maps identifies the FCHSD2 gene as an additional target of the enhancer cluster. CRISPR-Cas9-mediated deletion of the variant region, or of the associated enhancer, from human pancreas-derived EndoC-βH1 cells impairs glucose-stimulated insulin secretion. Expression of both STARD10 and FCHSD2 is reduced in cells harboring CRISPR deletions, and lower expression of STARD10 and FCHSD2 is associated, the latter nominally, with the possession of risk variant alleles in human islets. Finally, CRISPR-Cas9-mediated loss of STARD10 or FCHSD2, but not ARAP1, impairs regulated insulin secretion. Thus, multiple genes at the STARD10 locus influence β cell function.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Transferrin human, powder, BioReagent, suitable for cell culture
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Millipore
Nicotinamide, Precursor of the coenzymes NAD+ and NADP+.
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Rabbit IgG, Plasma
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®