Merck
  • Combination of Fish Oil and Selenium Enhances Anticancer Efficacy and Targets Multiple Signaling Pathways in Anti-VEGF Agent Treated-TNBC Tumor-Bearing Mice.
Combination of Fish Oil and Selenium Enhances Anticancer Efficacy and Targets Multiple Signaling Pathways in Anti-VEGF Agent Treated-TNBC Tumor-Bearing Mice.
Marine drugs (2021-04-04)
Chih-Hung Guo, Simon Hsia, Chieh-Han Chung, Yi-Chun Lin, Min-Yi Shih, Pei-Chung Chen, Guoo-Shyng W Hsu, Ciou-Ting Fan, Chia-Lin Peng
ABSTRACT
Fish oil (FO) and selenium (Se) possess antiangiogenic potential in malignant tumors. This study aimed to determine whether combination of FO and Se enhanced treatment efficacy of low-dose antiangiogenic agent Avastin (bevacizumab) in a dose-dependent manner and targeted multiple signaling pathways in triple-negative breast cancer (TNBC)-bearing mice. Randomized into five groups, mice received treatment with either physiological saline (control), Avastin alone, or Avastin in combination with low, medium, and high doses of FO/Se. The target signaling molecules for anticancer were determined either by measuring protein or mRNA expression. Avastin-treated mice receiving FO/Se showed lower tumor growth and metastasis than did mice treated with Avastin alone. Combination-treated mice exhibited lower expressions in multiple proangiogenic (growth) factors and their membrane receptors, and altered cytoplasmic signaling molecules (PI3K-PTEN-AKT-TSC-mTOR-p70S6K-4EBP1, Ras-Raf-MEK-ERK, c-Src-JAK2-STAT3-TMEPAI-Smad, LKB1-AMPK, and GSK3β/β-catenin). Dose-dependent inhibition of down-stream targets including epithelial-to-mesenchymal transition transcription factors, nuclear cyclin and cyclin-dependent kinases, cancer stem cell markers, heat shock protein (HSP-90), hypoxia-inducible factors (HIF-1α/-2α), matrix metalloprotease (MMP-9), and increased apoptosis were observed. These results suggest that combination treatment with FO and Se increases the therapeutic efficacy of Avastin against TNBC in a dose-dependent manner through multiple signaling pathways in membrane, cytoplasmic, and nucleic targets.
Sigma-Aldrich
Anti-CD44v10 Antibody, exon v10, Chemicon®, from rabbit
Sigma-Aldrich
Anti-C-X-C Chemokine Receptor 4 Antibody, extracellular loop, Chemicon®, from rabbit