Merck
  • Vitamin D suppresses bleomycin-induced pulmonary fibrosis by targeting the local renin-angiotensin system in the lung.
Vitamin D suppresses bleomycin-induced pulmonary fibrosis by targeting the local renin-angiotensin system in the lung.
Scientific reports (2021-08-18)
Jianjun Chang, Hongguang Nie, Xin Ge, Jie Du, Weicheng Liu, Xue Li, Yue Sun, Xinzhi Wei, Zhe Xun, Yan Chun Li
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a severe disorder leading to progressive and irreversible loss of pulmonary function. In this study we investigated the anti-fibrotic effect of vitamin D using a mouse model of IPF. Lung fibrosis was induced with bleomycin in vitamin D-sufficient and vitamin D-deficient C57BL/6 mice. We found that treatment with active vitamin D analog paricalcitol prevented mouse body weight loss and alleviated lung fibrosis, whereas vitamin D deficiency severely aggravated lung injury. At the molecular level, paricalcitol treatment suppressed the induction of fibrotic inducer TGF-β and extracellular matrix proteins α-SMA, collagen type I and fibronectin in the lung, whereas vitamin D deficiency exacerbated the induction of these proteins. Interestingly, bleomycin treatment activated the local renin-angiotensin system (RAS) in the lung, manifested by the induction of renin, angiotensinogen, angiotensin II and angiotensin receptor type 1 (AT1R). Paricalcitol treatment suppressed the induction of these RAS components, whereas vitamin D deficiency enhanced the activation of the lung RAS. We also showed that treatment of bleomycin-induced vitamin D-deficient mice with AT1R antagonist losartan relieved weight loss, substantially ameliorated lung fibrosis and markedly blocked TGF-β induction in the lung. Moreover, we demonstrated that in lung fibroblast cultures, TGF-β and angiotensin II synergistically induced TGF-β, AT1R, α-SMA, collagen type I and fibronectin, whereas 1,25-dihydroxyvitamin D markedly suppressed the induction of these fibrotic markers. Collectively, these observations strongly suggest that vitamin D mitigates lung fibrosis by blocking the activation of the lung RAS in this mouse model of IPF.
Sigma-Aldrich
Anti-Actin Antibody, smooth muscle, clone ASM-1, clone ASM-1, Chemicon®, from mouse