Merck
  • A Surface Plasmon Resonance Plastic Optical Fiber Biosensor for the Detection of Pancreatic Amylase in Surgically-Placed Drain Effluent.
A Surface Plasmon Resonance Plastic Optical Fiber Biosensor for the Detection of Pancreatic Amylase in Surgically-Placed Drain Effluent.
Sensors (Basel, Switzerland) (2021-06-03)
Laura Pasquardini, Nunzio Cennamo, Giuseppe Malleo, Lia Vanzetti, Luigi Zeni, Deborah Bonamini, Roberto Salvia, Claudio Bassi, Alessandra Maria Bossi
ABSTRACT
Postoperative pancreatic fistula (POPF), the major driver of morbidity and mortality following pancreatectomy, is caused by an abnormal communication between the pancreatic ductal epithelium and another epithelial surface containing pancreas-derived, enzyme-rich fluid. There is a strong correlation between the amylase content in surgically-placed drains early in the postoperative course and the development of POPF. A simple and cheap method to determine the amylase content from the drain effluent has been eagerly advocated. Here, we developed an amylase optical biosensor, based on a surface plasmon resonance (SPR) plastic optical fiber (POF), metallized with a 60 nm layer of gold and interrogated with white light. The sensor was made specific by coupling it with an anti-amylase antibody. Each surface derivatization step was optimized and studied by XPS, contact angle, and fluorescence. The POF-biosensor was tested for its response to amylase in diluted drain effluents. The volume of sample required was 50 µL and the measurement time was 8 min. The POF-biosensor showed selectivity for amylase, a calibration curve log-linear in the range of 0.8-25.8 U/L and a limit of detection (LOD) of ~0.5 U/L. In preliminary tests, the POF-biosensor allowed for the measurement of the amylase content of diluted surgically-placed drain effluents with an accuracy of >92% with respect to the gold standard. The POF-biosensor allows for reliable measurement and could be implemented to allow for a rapid bedside assessment of amylase value in drains following pancreatectomy.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution