Merck
  • A Murine Model of Chronic Lymphocytic Leukemia Based on B Cell-Restricted Expression of Sf3b1 Mutation and Atm Deletion.
A Murine Model of Chronic Lymphocytic Leukemia Based on B Cell-Restricted Expression of Sf3b1 Mutation and Atm Deletion.
Cancer cell (2019-02-05)
Shanye Yin, Rutendo G Gambe, Jing Sun, Aina Zurita Martinez, Zachary J Cartun, Fara Faye D Regis, Youzhong Wan, Jean Fan, Angela N Brooks, Sarah E M Herman, Elisa Ten Hacken, Amaro Taylor-Weiner, Laura Z Rassenti, Emanuela M Ghia, Thomas J Kipps, Esther A Obeng, Carrie L Cibulskis, Donna Neuberg, Dean R Campagna, Mark D Fleming, Benjamin L Ebert, Adrian Wiestner, Ignaty Leshchiner, James A DeCaprio, Gad Getz, Robin Reed, Ruben D Carrasco, Catherine J Wu, Lili Wang
ABSTRACT
SF3B1 is recurrently mutated in chronic lymphocytic leukemia (CLL), but its role in the pathogenesis of CLL remains elusive. Here, we show that conditional expression of Sf3b1-K700E mutation in mouse B cells disrupts pre-mRNA splicing, alters cell development, and induces a state of cellular senescence. Combination with Atm deletion leads to the overcoming of cellular senescence and the development of CLL-like disease in elderly mice. These CLL-like cells show genome instability and dysregulation of multiple CLL-associated cellular processes, including deregulated B cell receptor signaling, which we also identified in human CLL cases. Notably, human CLLs harboring SF3B1 mutations exhibit altered response to BTK inhibition. Our murine model of CLL thus provides insights into human CLL disease mechanisms and treatment.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction