Merck
  • Absence of the CXCR4 antagonist EPI-X4 from pharmaceutical human serum albumin preparations.
Absence of the CXCR4 antagonist EPI-X4 from pharmaceutical human serum albumin preparations.
Journal of translational medicine (2021-05-05)
Andrea Gilg, Mirja Harms, Lia-Raluca Olari, Ann-Kathrin Urbanowitz, Halvard Bonig, Jan Münch
ABSTRACT
Endogenous Peptide Inhibitor of CXCR4 (EPI-X4) is a natural antagonist of the CXC chemokine receptor 4 (CXCR4). EPI-X4 is a 16-mer peptide that is released from human serum albumin (HSA) by acidic aspartic proteases such as Cathepsin D and E. Since human serum albumin (HSA) is an important medicinal substance we asked whether different pharmaceutical HSA products contain EPI-X4 which could have been generated during manufacturing and whether HSA can serve as a substrate for cathepsins despite of the presence of stabilizers like caprylate. Eight pharmaceutical HSA preparations representing all currently used fractionation technologies were analyzed. The previously described specific EPI-X4 ELISA was used for quantification; in vitro EPI-X4 generation by acidification in the presence or absence of cathepsins was followed by quantification with ELISA. None of the pharmaceutical HSA preparations tested contained EPI-X4. Acidification of HSA did not generate EPI-X4. Addition of cathepsins D and E to acidified HSA yielded high concentrations of EPI-X4 in all HSA preparations, indistinguishable between individual products. Medicinal HSA preparations per se do not contain EPI-X4, but will replenish its precursor which can be cleaved to EPI-X4 in vivo, environmental conditions permitting.