Merck
PICK1 Deficiency Exacerbates Sepsis-Associated Acute Kidney Injury.
BioMed research international (2021-07-27)
Qian Dou, Hang Tong, Yichun Yang, Han Zhang, Hua Gan
ABSTRACT
We performed in vitro and in vivo experiments to explore the role of protein kinase C-binding protein 1 (PICK1), an intracellular transporter involved in oxidative stress-related neuronal diseases, in sepsis-related acute kidney injury (AKI). Firstly, PCR, western blotting, and immunohistochemistry were used to observe the expression of PICK1 after lipopolysaccharide- (LPS-) induced AKI. Secondly, by inhibiting PICK1 in vivo and silencing PICK1 in vitro, we further explored the effect of PICK1 on AKI. Finally, the relationship between PICK1 and oxidative stress and the related mechanisms were explored. We found that the expression of PICK1 was increased in LPS-induced AKI models both in vitro and in vivo. PICK1 silencing significantly aggravated LPS-induced apoptosis, accompanied by ROS production in renal tubular epithelial cells. FSC231, a PICK1-specific inhibitor, aggravated LPS-induced kidney injury. Besides, NAC (N-acetylcysteine), a potent ROS scavenger, significantly inhibited the PICK1-silencing-induced apoptosis. In conclusion, PICK1 might protect renal tubular epithelial cells from LPS-induced apoptosis by reducing excessive ROS, making PICK1 a promising preventive target in LPS-induced AKI.