Merck
  • Runx2 (Runt-Related Transcription Factor 2) Links the DNA Damage Response to Osteogenic Reprogramming and Apoptosis of Vascular Smooth Muscle Cells.
Runx2 (Runt-Related Transcription Factor 2) Links the DNA Damage Response to Osteogenic Reprogramming and Apoptosis of Vascular Smooth Muscle Cells.
Arteriosclerosis, thrombosis, and vascular biology (2020-12-29)
Andrew M Cobb, Syabira Yusoff, Robert Hayward, Sadia Ahmad, Mengxi Sun, Anja Verhulst, Patrick C D'Haese, Catherine M Shanahan
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
m-Aminophenylboronic acid–Agarose, aqueous suspension
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid