Merck
  • Design and diversity-oriented synthesis of novel 1,4-thiazepan-3-ones fused with bioactive heterocyclic skeletons and evaluation of their antioxidant and cytotoxic activities.
Design and diversity-oriented synthesis of novel 1,4-thiazepan-3-ones fused with bioactive heterocyclic skeletons and evaluation of their antioxidant and cytotoxic activities.
Bioorganic & medicinal chemistry letters (2011-12-06)
Feng Shi, Xiao-Ning Zeng, Xu-Dong Cao, Shu Zhang, Bo Jiang, Wei-Fa Zheng, Shu-Jiang Tu
ABSTRACT
This study has achieved the design and diversity-oriented synthesis of novel 1,4-thiazepine derivatives embedded with carbazole, pyrazole or isoxazole motif via microwave-assisted multicomponent reactions under solvent-free condition, thus providing a green and facile access to 1,4-thiazepine derivatives with prominent features of high structural diversity, short reaction time, high yields and environmental friendliness. More importantly, these novel compounds have been subjected to the test of in vitro antioxidant and cytotoxic activities, resulting in the finding that these 1,4-thiazepine derivatives not only have significant antioxidant activity, but also exhibit remarkably selective cytotoxicity to carcinoma cell line HCT 116.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)