Merck
  • Creb5 establishes the competence for Prg4 expression in articular cartilage.
Creb5 establishes the competence for Prg4 expression in articular cartilage.
Communications biology (2021-03-14)
Cheng-Hai Zhang, Yao Gao, Unmesh Jadhav, Han-Hwa Hung, Kristina M Holton, Alan J Grodzinsky, Ramesh A Shivdasani, Andrew B Lassar
ABSTRACT
A hallmark of cells comprising the superficial zone of articular cartilage is their expression of lubricin, encoded by the Prg4 gene, that lubricates the joint and protects against the development of arthritis. Here, we identify Creb5 as a transcription factor that is specifically expressed in superficial zone articular chondrocytes and is required for TGF-β and EGFR signaling to induce Prg4 expression. Notably, forced expression of Creb5 in chondrocytes derived from the deep zone of the articular cartilage confers the competence for TGF-β and EGFR signals to induce Prg4 expression. Chromatin-IP and ATAC-Seq analyses have revealed that Creb5 directly binds to two Prg4 promoter-proximal regulatory elements, that display an open chromatin conformation specifically in superficial zone articular chondrocytes; and which work in combination with a more distal regulatory element to drive induction of Prg4 by TGF-β. Our results indicate that Creb5 is a critical regulator of Prg4/lubricin expression in the articular cartilage.
Sigma-Aldrich
Anti-Lubricin Antibody/Proteoglycan 4, clone 9G3, clone 9G3, from mouse
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Roche
Anti-Fluorescein-POD, Fab fragments, from sheep
Roche
Fluorescein RNA Labeling Mix, solution, suitable for Northern blotting, suitable for Southern blotting, suitable for hybridization