Merck
  • The pluripotent stem cell-specific transcript ESRG is dispensable for human pluripotency.
The pluripotent stem cell-specific transcript ESRG is dispensable for human pluripotency.
PLoS genetics (2021-05-26)
Kazutoshi Takahashi, Michiko Nakamura, Chikako Okubo, Zane Kliesmete, Mari Ohnuki, Megumi Narita, Akira Watanabe, Mai Ueda, Yasuhiro Takashima, Ines Hellmann, Shinya Yamanaka
ABSTRACT
Human pluripotent stem cells (PSCs) express human endogenous retrovirus type-H (HERV-H), which exists as more than a thousand copies on the human genome and frequently produces chimeric transcripts as long-non-coding RNAs (lncRNAs) fused with downstream neighbor genes. Previous studies showed that HERV-H expression is required for the maintenance of PSC identity, and aberrant HERV-H expression attenuates neural differentiation potentials, however, little is known about the actual of function of HERV-H. In this study, we focused on ESRG, which is known as a PSC-related HERV-H-driven lncRNA. The global transcriptome data of various tissues and cell lines and quantitative expression analysis of PSCs showed that ESRG expression is much higher than other HERV-Hs and tightly silenced after differentiation. However, the loss of function by the complete excision of the entire ESRG gene body using a CRISPR/Cas9 platform revealed that ESRG is dispensable for the maintenance of the primed and naïve pluripotent states. The loss of ESRG hardly affected the global gene expression of PSCs or the differentiation potential toward trilineage. Differentiated cells derived from ESRG-deficient PSCs retained the potential to be reprogrammed into induced PSCs (iPSCs) by the forced expression of OCT3/4, SOX2, and KLF4. In conclusion, ESRG is dispensable for the maintenance and recapturing of human pluripotency.
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin, smooth muscle Antibody, clone ASM-1/1A4, clone ASM-1 (1A4), from mouse