Merck
  • Src-IL-18 signaling regulates the secretion of atrial natriuretic factor in hypoxic beating rat atria.
Src-IL-18 signaling regulates the secretion of atrial natriuretic factor in hypoxic beating rat atria.
Kardiologia polska (2021-06-28)
Xiang Li, Cheng-Xi Wei, Cheng-Zhe Wu, Lan Hong, Zhuo-Na Han, Ying Liu, Wang Yue-Ying, Xun Cui
ABSTRACT
Interleukin (IL)-18 is produced mainly in the heart and can be associated with the development of cardiac hypertrophy that leads to cardiac dysfunction. However, the effects of hypoxia on IL-18 expression and atrial natriuretic factor (ANF) secretion remain largely unknown. The aim of this study was to assess the effect of hypoxia on IL-18 production and its role in ANF secretion by using an isolated perfused beating rat atrial model. The level of ANF in the perfusates was determined by radioimmunoassay, and the protein levels of Src, IL-18 and its receptors (IL-18-Rα and IL-18-Rβ), Rho guanine nucleotide exchange factor (RhoGEF) and RhoA, activating transcription factor 3 (ATF3), T cell factor (TCF) 3 and 4, and lymphoid enhancer factor (LEF) 1 in atrial tissue samples were detected by Western blotting. Hypoxia significantly upregulated the expression of the non-receptor tyrosine kinase Src, and this effect was blocked by endothelin-1 receptor type A (BQ123) and type B (BQ788) antagonists. Hypoxia also enhanced the expression of RhoGEF and RhoA concomitantly with the upregulation of IL-18, IL-18-Rα and IL-18-Rβ. The hypoxia-induced RhoGEF and RhoA were abolished by Src inhibitor 1 (SrcI), and the protein levels of IL-18 and its two receptors were also blocked by SrcI. Moreover, the hypoxia-induced expression levels of ATF3, TCF3, TCF4 and LEF1 were repealed by IL-18 binding protein, and the hypoxia-promoted secretion of ANF was also obviously attenuated by this binding protein. These findings imply that Src-IL-18 signaling is involved in the release of ANF in hypoxic beating rat atria.
Sigma-Aldrich
Anti-Src Antibody, clone N6L, rabbit monoclonal, culture supernatant, clone N6L, from rabbit
Sigma-Aldrich
BQ-788, ≥95%, solid