Merck
  • Receptor-mediated mitophagy regulates EPO production and protects against renal anemia.
Receptor-mediated mitophagy regulates EPO production and protects against renal anemia.
eLife (2021-05-05)
Guangfeng Geng, Jinhua Liu, Changlu Xu, Yandong Pei, Linbo Chen, Chenglong Mu, Ding Wang, Jie Gao, Yue Li, Jing Liang, Tian Zhao, Chuanmei Zhang, Jiaxi Zhou, Quan Chen, Yushan Zhu, Lihong Shi
ABSTRACT
Erythropoietin (EPO) drives erythropoiesis and is secreted mainly by the kidney upon hypoxic or anemic stress. The paucity of EPO production in renal EPO-producing cells (REPs) causes renal anemia, one of the most common complications of chronic nephropathies. Although mitochondrial dysfunction is commonly observed in several renal and hematopoietic disorders, the mechanism by which mitochondrial quality control impacts renal anemia remains elusive. In this study, we showed that FUNDC1, a mitophagy receptor, plays a critical role in EPO-driven erythropoiesis induced by stresses. Mechanistically, EPO production is impaired in REPs in Fundc1-/- mice upon stresses, and the impairment is caused by the accumulation of damaged mitochondria, which consequently leads to the elevation of the reactive oxygen species (ROS) level and triggers inflammatory responses by up-regulating proinflammatory cytokines. These inflammatory factors promote the myofibroblastic transformation of REPs, resulting in the reduction of EPO production. We therefore provide a link between aberrant mitophagy and deficient EPO generation in renal anemia. Our results also suggest that the mitochondrial quality control safeguards REPs under stresses, which may serve as a potential therapeutic strategy for the treatment of renal anemia.
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, liquid, sterile-filtered, With sodium bicarbonate, without L-glutamine, suitable for cell culture, suitable for hybridoma
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Bovine Serum Albumin solution, 10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered