Merck
  • Prediction of drug intestinal absorption by new linear and non-linear QSPR.
Prediction of drug intestinal absorption by new linear and non-linear QSPR.
European journal of medicinal chemistry (2010-11-30)
Alan Talevi, Mohammad Goodarzi, Erlinda V Ortiz, Pablo R Duchowicz, Carolina L Bellera, Guido Pesce, Eduardo A Castro, Luis E Bruno-Blanch
ABSTRACT
In order to minimize the high attrition rate that usually characterizes drug research and development projects, current medicinal chemists aim to characterize both pharmacological and ADME profiles at the beginning of drug R&D initiatives. Thus, the development of ADME High-Throughput Screening in vitro and in silico ADME models has become an important growing research area. Here we present new linear and non-linear predictive QSPR models to predict the human intestinal absorption rate, which are derived from a medium sized, balanced and diverse training set of organic compounds. The structure-property relationships so obtained involve only 4 molecular descriptors, and display an excellent ratio of number of cases to number of descriptors. Their adjustment of the training set data together with the performance achieved during the internal and external validation procedures are comparable to previously reported modeling efforts.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Ibuprofen
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Warfarin, PESTANAL®, analytical standard
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
4-Dimethylaminoantipyrine, reactive nitrogen species scavenger
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture