Merck
Microglia control small vessel calcification via TREM2.
Science advances (2021-02-28)
Yvette Zarb, Sucheta Sridhar, Sina Nassiri, Sebastian Guido Utz, Johanna Schaffenrath, Upasana Maheshwari, Elisabeth J Rushing, K Peter R Nilsson, Mauro Delorenzi, Marco Colonna, Melanie Greter, Annika Keller
ABSTRACT
Microglia participate in central nervous system (CNS) development and homeostasis and are often implicated in modulating disease processes. However, less is known about the role of microglia in the biology of the neurovascular unit (NVU). In particular, data are scant on whether microglia are involved in CNS vascular pathology. In this study, we use a mouse model of primary familial brain calcification, Pdgfbret/ret , to investigate the role of microglia in calcification of the NVU. We report that microglia enclosing vessel calcifications, coined calcification-associated microglia, display a distinct activation phenotype. Pharmacological ablation of microglia with the CSF1R inhibitor PLX5622 leads to aggravated vessel calcification. Mechanistically, we show that microglia require functional TREM2 for controlling vascular calcification. Our results demonstrate that microglial activity in the setting of pathological vascular calcification is beneficial. In addition, we identify a previously unrecognized function of microglia in halting the expansion of vascular calcification.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
5-Ethynyl-2′-deoxyuridine, 95%
Sigma-Aldrich
Tamoxifen, ≥99%