Merck
  • A-CD estrogens. I. Substituent effects, hormone potency, and receptor subtype selectivity in a new family of flexible estrogenic compounds.
A-CD estrogens. I. Substituent effects, hormone potency, and receptor subtype selectivity in a new family of flexible estrogenic compounds.
Journal of medicinal chemistry (2010-12-31)
James S Wright, Hooman Shadnia, James M Anderson, Tony Durst, Muhammad Asim, Mohamed El-Salfiti, Christine Choueiri, M A Christine Pratt, Samantha C Ruddy, Rosanna Lau, Kathryn E Carlson, John A Katzenellenbogen, Peter J O'Brien, Luke Wan
ABSTRACT
Long-term use of estrogen supplements by women leads to an increased risk of breast and uterine cancers. Possible mechanisms include metabolism of estradiol and compounds related to tumor-initiating quinones, and ligand-induced activation of the estrogen receptors ERα and ERβ which can cause cancer cell proliferation, depending on the ratio of receptors present. One therapeutic goal would be to create a spectrum of compounds of variable potency for ERα and ERβ, which are resistant to quinone formation, and to determine an optimum point in this spectrum. We describe the synthesis, modeling, binding affinities, hormone potency, and a measure of quinone formation for a new family of A-CD estrogens, where the A-C bond is formed by ring coupling. Some substituents on the A-ring increase hormone potency, and one compound is much less quinone-forming than estradiol. These compounds span a wide range of receptor subtype selectivities and may be useful in hormone replacement therapy.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications