Merck
  • JMJD6 Is a Druggable Oxygenase That Regulates AR-V7 Expression in Prostate Cancer.
JMJD6 Is a Druggable Oxygenase That Regulates AR-V7 Expression in Prostate Cancer.
Cancer research (2021-04-07)
Alec Paschalis, Jonathan Welti, Antje J Neeb, Wei Yuan, Ines Figueiredo, Rita Pereira, Ana Ferreira, Ruth Riisnaes, Daniel Nava Rodrigues, Juan M Jiménez-Vacas, Soojin Kim, Takuma Uo, Patrizio Di Micco, Anthony Tumber, Md Saiful Islam, Marc A Moesser, Martine Abboud, Akane Kawamura, Bora Gurel, Rossitza Christova, Veronica S Gil, Lorenzo Buroni, Mateus Crespo, Susana Miranda, Maryou B Lambros, Suzanne Carreira, Nina Tunariu, Andrea Alimonti, Bissan Al-Lazikani, Christopher J Schofield, Stephen R Plymate, Adam Sharp, Johann S de Bono
ABSTRACT
Endocrine resistance (EnR) in advanced prostate cancer is fatal. EnR can be mediated by androgen receptor (AR) splice variants, with AR splice variant 7 (AR-V7) arguably the most clinically important variant. In this study, we determined proteins key to generating AR-V7, validated our findings using clinical samples, and studied splicing regulatory mechanisms in prostate cancer models. Triangulation studies identified JMJD6 as a key regulator of AR-V7, as evidenced by its upregulation with in vitro EnR, its downregulation alongside AR-V7 by bromodomain inhibition, and its identification as a top hit of a targeted siRNA screen of spliceosome-related genes. JMJD6 protein levels increased (P < 0.001) with castration resistance and were associated with higher AR-V7 levels and shorter survival (P = 0.048). JMJD6 knockdown reduced prostate cancer cell growth, AR-V7 levels, and recruitment of U2AF65 to AR pre-mRNA. Mutagenesis studies suggested that JMJD6 activity is key to the generation of AR-V7, with the catalytic machinery residing within a druggable pocket. Taken together, these data highlight the relationship between JMJD6 and AR-V7 in advanced prostate cancer and support further evaluation of JMJD6 as a therapeutic target in this disease. SIGNIFICANCE: This study identifies JMJD6 as being critical for the generation of AR-V7 in prostate cancer, where it may serve as a tractable target for therapeutic intervention.
Sigma-Aldrich
MISSION® esiRNA, targeting human JMJD6
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
2,4-Pyridinedicarboxylic acid, ≥98.0%
Sigma-Aldrich
EZ-Magna NuCLEAR RIP (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit, EZ-Magna Nuclear RIP (Cross-Linked) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ACS reagent, 99%