Merck
  • Longitudinal study of Pex1-G844D NMRI mouse model: A robust pre-clinical model for mild Zellweger spectrum disorder.
Longitudinal study of Pex1-G844D NMRI mouse model: A robust pre-clinical model for mild Zellweger spectrum disorder.
Biochimica et biophysica acta. Molecular basis of disease (2020-07-22)
Tanguy Demaret, Martin Roumain, Jérôme Ambroise, Jonathan Evraerts, Joachim Ravau, Caroline Bouzin, Bertrand Bearzatto, Jean-Luc Gala, Hedwig Stepman, Sandrine Marie, Marie-Françoise Vincent, Giulio G Muccioli, Mustapha Najimi, Etienne M Sokal
ABSTRACT
Zellweger spectrum disorders (ZSD) are inborn errors of metabolism caused by mutations in PEX genes that lead to peroxisomal biogenesis disorder (PBD). No validated treatment is able to modify the dismal progression of the disease. ZSD mouse models used to develop therapeutic approaches are limited by poor survival and breeding restrictions. To overcome these limitations, we backcrossed the hypomorphic Pex1 p.G844D allele to NMRI background. NMRI mouse breeding restored an autosomal recessive Mendelian inheritance pattern and delivered twice larger litters. Mice were longitudinally phenotyped up to 6 months of age to make this model suitable for therapeutic interventions. ZSD mice exhibited growth retardation and relative hepatomegaly associated to progressive hepatocyte hypertrophy. Biochemical studies associated with RNA sequencing deciphered ZSD liver glycogen metabolism alterations. Affected fibroblasts displayed classical immunofluorescence pattern and biochemical alterations associated with PBD. Plasma and liver showed very long-chain fatty acids, specific oxysterols and C27 bile acids intermediates elevation in ZSD mice along with a specific urine organic acid profile. With ageing, C26 fatty acid and phytanic acid levels tended to normalize in ZSD mice, as described in patients reaching adulthood. In conclusion, our mouse model recapitulates a mild ZSD phenotype and is suitable for liver-targeted therapies evaluation.
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
L-Pipecolic acid, 99% (titration)
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
DAPI, for nucleic acid staining