Merck
  • SCY-635, a novel nonimmunosuppressive analog of cyclosporine that exhibits potent inhibition of hepatitis C virus RNA replication in vitro.
SCY-635, a novel nonimmunosuppressive analog of cyclosporine that exhibits potent inhibition of hepatitis C virus RNA replication in vitro.
Antimicrobial agents and chemotherapy (2009-11-26)
Sam Hopkins, Bernard Scorneaux, Zhuhui Huang, Michael G Murray, Stephen Wring, Craig Smitley, Richard Harris, Frank Erdmann, Gunter Fischer, Yves Ribeill
ABSTRACT
SCY-635 is a novel nonimmunosuppressive cyclosporine-based analog that exhibits potent suppression of hepatitis C virus (HCV) replication in vitro. SCY-635 inhibited the peptidyl prolyl isomerase activity of cyclophilin A at nanomolar concentrations but showed no detectable inhibition of calcineurin phosphatase activity at concentrations up to 2 microM. Metabolic studies indicated that SCY-635 did not induce the major cytochrome P450 enzymes 1A2, 2B6, and 3A4. SCY-635 was a weak inhibitor and a poor substrate for P-glycoprotein. Functional assays with stimulated Jurkat cells and stimulated human peripheral blood mononuclear cells indicated that SCY-635 is a weaker inhibitor of interleukin-2 secretion than cyclosporine. A series of two-drug combination studies was performed in vitro. SCY-635 exhibited synergistic antiviral activity with alpha interferon 2b and additive antiviral activity with ribavirin. SCY-635 was shown to be orally bioavailable in multiple animal species and produced blood and liver concentrations of parent drug that exceeded the 50% effective dose determined in the bicistronic con1b-derived replicon assay. These results suggest that SCY-635 warrants further investigation as a novel therapeutic agent for the treatment of individuals who are chronically infected with HCV.
Product Description
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Warfarin, PESTANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
3-Methylcholanthrene, analytical standard
Sigma-Aldrich
3-Methylcholanthrene, 98%