Merck
  • ANKFY1 is essential for retinal endothelial cell proliferation and migration via VEGFR2/Akt/eNOS pathway.
ANKFY1 is essential for retinal endothelial cell proliferation and migration via VEGFR2/Akt/eNOS pathway.
Biochemical and biophysical research communications (2020-10-24)
Miruto Tanaka, Shinsuke Nakamura, Masashi Maekawa, Shigeki Higashiyama, Hideaki Hara
ABSTRACT
Dysregulation of endothelial cell proliferation and migration are hallmarks of angiogenic diseases. Among them, excessive ocular angiogenesis is a major cause of blindness. Vascular endothelial growth factor (VEGF)-VEGF receptor 2 (VEGFR2) signaling plays crucial roles in angiogenesis, endothelial cell proliferation and migration. Here, we showed that ankyrin repeat and FYVE domain containing 1 (ANKFY1), a Rab5-GTP-interacting protein, is required for retinal endothelial cell proliferation and migration. ANKFY1 knockdown significantly suppressed cell growth of human retinal microvascular endothelial cells (HRMECs) in the presence or absence of VEGF. HRMEC migration was also inhibited by depletion of ANKFY1. Western blot analysis showed that ANKFY1 knockdown reduced cell surface VEGFR2 level. In contrast, qRT-PCR analysis indicated that ANKFY1 knockdown had no effect on VEGFR2 mRNA levels. We also found that the attenuation of the protein kinase B/endothelial nitric oxide synthase (Akt/eNOS) pathway in ANKFY1 knockdown HRMECs. In conclusion, our findings revealed novel functions of ANKFY1 in cell growth and migration of retinal endothelial cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human ANKFY1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$31.70
25 g
In Stock
Details...
$38.80
100 g
In Stock
Details...
$120.00
1 kg
In Stock
Details...
$796.00