Merck
  • Polysulfated Hyaluronan GlycoMira-1111 Inhibits Elastase and Improves Rheology in Cystic Fibrosis Sputum.
Polysulfated Hyaluronan GlycoMira-1111 Inhibits Elastase and Improves Rheology in Cystic Fibrosis Sputum.
American journal of respiratory cell and molecular biology (2020-12-03)
Apparao B Kummarapurugu, Shuo Zheng, Abigail Pulsipher, Justin R Savage, Jonathan Ma, Bruce K Rubin, Thomas P Kennedy, Judith A Voynow
ABSTRACT
Cystic fibrosis (CF) lung disease is marked by high concentrations of neutrophil elastase (NE) and DNA polymers; both factors contribute to airway disease. Although inhaled recombinant human dornase alfa reduces the frequency of CF pulmonary exacerbations, it also increases free NE activity in the sputum. There are no approved anti-NE therapies for patients with CF. We investigated whether synthetic, low-molecular weight polysulfated hyaluronan GlycoMira-1111 (GM-1111) would be effective as an anti-NE drug using ex vivo CF sputum. Anti-NE activity of GM-1111 was tested in CF sputum in the presence or absence of dornase alfa and/or hypertonic saline using a spectrophotometric assay specific for human NE and was compared with unfractionated heparin. We tested whether GM-1111 disaggregated DNA from CF sputum (using gel electrophoresis analysis) or modified CF sputum viscoelastic properties (using a dynamic rheometer). GM-1111 and unfractionated heparin had near equivalent anti-NE activity in CF sputum in the presence of dornase alfa. Both GM-1111 and unfractionated heparin retained anti-NE activity in hypertonic saline but with decreased activity. GM-1111 increased the release of soluble DNA in CF sputum, resulting in improved depolymerization efficacy of dornase alfa. GM-1111 decreased CF sputum elasticity. GM-1111 inhibited NE activity, enhanced DNA depolymerization by deoxyribonuclease, and decreased viscoelastic properties of CF sputum, similar to effects reported previously for unfractionated heparin. Unlike heparins, GM-1111 is synthetic, with minimal anticoagulant activity, and is not derived from animal products. These key attributes provide advantages over unfractionated heparin as a potential therapeutic for CF.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein