Merck
  • Artificial switching of the metabolic processing pathway of an etiologic factor, β2-microglobulin, by a "navigator" molecule.
Artificial switching of the metabolic processing pathway of an etiologic factor, β2-microglobulin, by a "navigator" molecule.
Journal of controlled release : official journal of the Controlled Release Society (2020-08-04)
Yusuke Kambe, Kento Koyashiki, Yoshiaki Hirano, Mariko Harada-Shiba, Tetsuji Yamaoka
ABSTRACT
Metabolic pathways in the body are highly specific. Dysfunction of a metabolic pathway triggers the accumulation of its target substance. For example, kidney failure results in increased β2-microglobulin blood levels, causing dialysis-related amyloidosis. Previously, we proposed a novel therapeutic concept, that is a removal of an etiologic factor of metabolic disease by artificial switching of its metabolic processing pathway, and tested this concept using in cultured cells. However, the feasibility of artificial metabolic switching in vivo remained unknown. Here, we show that a newly developed "navigator" molecule changes the metabolic processing pathway of β2-microglobulin from the kidney to the liver in mouse. The artificial metabolic switching is achieved by the capture of the etiologic factor by the navigator, which then steers the etiologic factor to hepatic lysosomes via low-density lipoprotein receptors. These findings demonstrate that navigator-based artificial metabolic switching can be a therapeutic strategy for various diseases caused by metabolic disorders.