Merck
  • Yersinia YopT inhibits RLH-mediated NF-κB and IRF3 signal transduction.
Yersinia YopT inhibits RLH-mediated NF-κB and IRF3 signal transduction.
Microbiology and immunology (2020-09-10)
Dongyu Li, Haoyong Wang, Pingping Zhang, Yanhong Zhang, Xiang He, Hui Zhong, Kai Guan, Min Min, Qi Gao, Congwen Wei
ABSTRACT
The Gram-negative bacterial pathogen Yersinia delivers six effector proteins into the host cells to block the host innate immune response. One of the effectors, YopT, is a potent cysteine protease that causes the disruption of the actin cytoskeleton to inhibit phagocytosis of the pathogen; however, its molecular mechanism and relevance to pathogenesis need further investigation. In this report, we show that RIG-I is a novel target of the YopT protein. Remarkably, YopT interacts with RIG-I and inhibits rat liver homogenate-mediated nuclear factor-κB and interferon regulatory factor-3 activation. Further studies revealed a YopT-dependent increase in the K48-polymerized ubiquitination of RIG-I. These findings suggest that YopT negatively regulates RIG-I-mediated cellular antibacterial response by targeting RIG-I.
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid