Merck
  • Effect of UV/chlorine treatment on photophysical and photochemical properties of dissolved organic matter.
Effect of UV/chlorine treatment on photophysical and photochemical properties of dissolved organic matter.
Water research (2021-02-01)
Yangjian Zhou, Fangyuan Cheng, Dongyang He, Ya-Nan Zhang, Jiao Qu, Xin Yang, Jingwen Chen, Willie J G M Peijnenburg
ABSTRACT
Dissolved organic matter (DOM) is a ubiquitous component in effluents, DOM discharged with an effluent can affect the composition and properties of natural DOM in the receiving waters. As the photophysical and photochemical properties of effluent DOM can be changed by wastewater treatment processes, the effect of UV/chlorine treatment on the photophysical and photochemical properties of DOM was investigated using Suwannee River fulvic acid (SRFA) and Suwannee River natural organic matter (SRNOM) as representatives. Results showed that the absorbance of the two DOM was significantly decreased. The evolution trends of three representative photophysical parameters upon increase of chlorine dosages were observed. Also, a decrease in DOM aromaticity, molecular weight and electron-donating capacity was observed upon increasing chlorine dosage. Quantum yields of excited triplet state of DOM (3DOM*), singlet oxygen (1O2) and hydroxyl radicals (·OH) first decreases and then increased in the UV/chlorine systems upon increasing chlorine dosages due to the different reaction pathways of the two DOM. Moreover, 3DOM* can not only be regarded as a "controller" of other reactive intermediates, but also effectively promote the photodegradation of bezafibrate, which is classified as a persistent organic contaminant. This study gives deep insights into effects of UV/chlorine on the photophysical and photochemical properties of DOM, and is helpful for understanding the dynamic roles of DOM in the photodegradation of micropollutants.
Sigma-Aldrich
Chlorohydroquinone, technical grade, 85%
Sigma-Aldrich
Thymidine 5′-monophosphate disodium salt hydrate, ≥99%
Sigma-Aldrich
4-Hydroxybenzophenone, 98%