Merck
  • C-Type Lectin Maintains the Homeostasis of Intestinal Microbiota and Mediates Biofilm Formation by Intestinal Bacteria in Shrimp.
C-Type Lectin Maintains the Homeostasis of Intestinal Microbiota and Mediates Biofilm Formation by Intestinal Bacteria in Shrimp.
Journal of immunology (Baltimore, Md. : 1950) (2021-02-03)
Yu-Xuan Zhang, Ming-Lu Zhang, Xian-Wei Wang
ABSTRACT
Intestinal microbiota are closely related to host physiology. Over the long course of evolution and interaction, both commensal bacteria and their host have evolved multiple strategies to adapt to each other. However, in invertebrates, the regulatory mechanism of intestinal microbiota homeostasis is largely unknown. In the current study, a digestive tract-specific C-type lectin, designated as CTL33, was identified because of its abundance and response to bacteria in the intestine of kuruma shrimp (Marsupenaeus japonicus). Silencing of CTL33 expression led directly to intestinal dysbiosis, tissue damage, and shrimp death. CTL33 could facilitate biofilm formation by the intestinal bacteria. This function originated from its unique architecture, with a lectin domain responsible for bacteria recognition and a coiled coil region that mediated CTL33 dimerization and cross-linked the bacteria into a biofilm-like complex. By mediating the formation of a biofilm, CTL33 promoted the establishment of intestinal bacteria in intestine and maintained the homeostasis of the microbiota. Thus, to our knowledge, we demonstrated a new mechanism of C-type lectin-mediated biofilm formation by intestinal bacteria, providing new insights into intestinal homeostasis regulation in invertebrates.
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Peptidoglycan from Staphylococcus aureus, cell wall component