Merck
  • Reliability of bond dissociation enthalpy calculated by the PM6 method and experimental TEAC values in antiradical QSAR of flavonoids.
Reliability of bond dissociation enthalpy calculated by the PM6 method and experimental TEAC values in antiradical QSAR of flavonoids.
Bioorganic & medicinal chemistry (2009-12-01)
Dragan Amić, Bono Lucić
ABSTRACT
The applicability of the newly developed RM1 and PM6 methods implemented in the semiempirical quantum chemistry mopac2009 software package in modeling free radical scavenging activity of flavonoids was examined. Bond dissociation enthalpy (BDE) of OH groups could be calculated much faster than with DFT method but with similar quality. Despite the known shortcomings of the Trolox equivalent antioxidant capacity (TEAC) assay, we show that taking into account the hydrogen atom transfer (HAT) mechanism of free radical scavenging of flavonoids encoded by minimal BDE values (BDE(min)) and the number of OH groups (nOH), as well as experimental data, reasonable QSAR models could be developed. For TEAC values of 38 flavonoids measured by the ABTS free radical, a model based on BDE(min) and nOH was developed, having very good statistical parameters (r=0.983, r(cv)=0.976). The applicability of this model to three different data sets of flavonoids and reliability of TEAC values measured in distinct laboratories were discussed. Finally, a reasonably good model of experimental vitamin C equivalent antioxidant capacity (VCEAC) of 36 flavonoids was obtained (r=0.954, r(cv)=0.947), involving BDE(min) and nOH as descriptors. Additionally, all presented models have comparable fit and cross-validated statistical parameters, as well as significant regression coefficients.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Pelargonidin chloride
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Supelco
(−)-Epigallocatechin, analytical standard
Supelco
Procyanidin B2, analytical standard
Sigma-Aldrich
Idaein chloride, ≥90% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)